Versilon® PFA Tubing
Exhibits superior chemical and diffusion resistance
  • High-purity grade fluoropolymer—ideal for extremely critical applications
  • Enhanced burst strength
  • Withstands up to 206°C (500°F)
  • Retains excellent mechanical strength even at elevated temperatures
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
Made of high-purity grade fluoropolymer, Versilon® PFA tubing provides superior chemical and diffusion resistance. Use in critical applications where precise quality control is required, such as semi-conductor and environmental industries. Tubing maintains great mechanical strength and resists stress cracking even at high temperatures.
$152.25 - $1,930.00USD / Pkg of 1
17 variations of this product are available.
See All Product Options Below
17 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
Versilon PFA Tubing, 1/16" ID x 1/8"OD; 50 Ft EW-50107-39 Mfr # TSPF35-0125-031-50
COMPARE
1/16 1/8 1/32
$152.25USD / Pkg of 1
Versilon PFA Tubing, 1/8" ID x 3/16"OD; 50 Ft EW-50107-40 Mfr # TSPF35-0187-031-50
COMPARE
1/8 3/16 1/32
$252.00USD / Pkg of 1
Versilon PFA Tubing, 3/16" ID x 1/4"OD; 50 Ft EW-50107-41 Mfr # TSPF35-0250-031-50
COMPARE
3/16 1/4 1/32
$325.50USD / Pkg of 1
Versilon PFA Tubing, 5/32" ID x 1/4"OD; 50 Ft EW-50107-42 Mfr # TSPF35-0250-047-50
COMPARE
5/32 1/4 3/64
$467.50USD / Pkg of 1
Versilon PFA Tubing, 1/8" ID x 1/4"OD; 50 Ft EW-50107-43 Mfr # TSPF35-0250-062-50
COMPARE
1/8 1/4 1/16
$552.00USD / Pkg of 1
Versilon PFA Tubing, 1/4" ID x 5/16"OD; 50 Ft EW-50107-44 Mfr # TSPF35-0312-031-50
COMPARE
1/4 5/16 1/32
$420.00USD / Pkg of 1
Versilon PFA Tubing, 3/16" ID x 5/16"OD; 50 Ft EW-50107-45 Mfr # TSPF35-0312-062-50
COMPARE
3/16 5/16 1/16
$760.00USD / Pkg of 1
Versilon PFA Tubing, 5/16" ID x 3/8"OD; 50 Ft EW-50107-46 Mfr # TSPF35-0375-031-50
COMPARE
5/16 3/8 1/32
$525.00USD / Pkg of 1
Versilon PFA Tubing, 1/4" ID x 3/8"OD; 50 Ft EW-50107-47 Mfr # TSPF35-0375-062-50
COMPARE
1/4 3/8 1/16
$843.00USD / Pkg of 1
Versilon PFA Tubing, 3/8" ID x 7/16"OD; 50 Ft EW-50107-48 Mfr # TSPF35-0437-031-50
COMPARE
3/8 7/16 1/32
$630.00USD / Pkg of 1
Versilon PFA Tubing, 7/16" ID x 1/2"OD; 50 Ft EW-50107-49 Mfr # TSPF35-0500-031-50
COMPARE
7/16 1/2 1/32
$713.00USD / Pkg of 1
Versilon PFA Tubing, 3/8" ID x 1/2"OD; 50 Ft EW-50107-50 Mfr # TSPF35-0500-062-50
COMPARE
3/8 1/2 1/16
$1,045.00USD / Pkg of 1
Versilon PFA Tubing, 1/2" ID x 9/16"OD; 50 Ft EW-50107-51 Mfr # TSPF35-0562-031-50
COMPARE
1/2 9/16 1/32
$796.00USD / Pkg of 1
Versilon PFA Tubing, 1/2" ID x 5/8"OD; 50 Ft EW-50107-52 Mfr # TSPF35-0625-062-50
COMPARE
1/2 5/8 1/16
$1,240.00USD / Pkg of 1
Versilon PFA Tubing, 5/8" ID x 3/4"OD; 50 Ft EW-50107-53 Mfr # TSPF35-0750-062-50
COMPARE
5/8 3/4 1/16
$1,615.00USD / Pkg of 1
Versilon PFA Tubing, 3/4" ID x 7/8"OD; 50 Ft EW-50107-54 Mfr # TSPF35-0875-062-50
COMPARE
3/4 7/8 1/16
$1,930.00USD / Pkg of 1
Versilon PFA Tubing, 7/8" ID x 1"OD; 50 Ft EW-50107-55 Mfr # TSPF35-1000-062-50
COMPARE
7/8 1 1/16
$1,530.00USD / Pkg of 1
Showing 1- 17 of 17