VistaLab™ VistaRak™ Pipette Tips
Maximize performance and benefits of your pipette
  • Tips made of 100% pure virgin polypropylene
  • Save space—192 tips per rack with a footprint of just 4.5” x 6”
VistaRak™ polypropylene (PP) pipette tips contain 192 tips in convenient 16 x 12 multichannel layout in a hinged box with a small footprint of just 4.5” x 6”. Each model contains 5 racks of 192 tips. Tips are compatible with VistaLab™ Ovation and MLA model pipettors.
$89.90 - $242.00USD / Case of 960
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (µL) Sterile Tip Style
Availability Pricing
VistaLab VistaRak™ Pipette Tips, Non-Sterile, 25 µL; 192 Tips x 5 Racks EW-06197-70 Mfr # 4060-1002
COMPARE
25 No Micro; Standard
$91.50USD / Case of 960
VistaLab VistaRak™ Pipette Tips, Non-Sterile, 250 µL; 192 Tips x 5 Racks EW-06197-71 Mfr # 4060-2004
COMPARE
250 No Small; Standard
$89.90USD / Case of 960
VistaLab VistaRak™ Pipette Tips, Non-Sterile, 1250 µL; 192 Tips x 5 Racks EW-06197-72 Mfr # 4060-3004
COMPARE
1250 No Large; Standard
$111.75USD / Case of 960
VistaLab VistaRak™ Pipette Tips, Sterile, RNase/DNase Free, 25 µL; 192 Tips x 5 Racks EW-06197-76 Mfr # 4060-1032
COMPARE
25 Yes Micro; RNase/DNase Free
$112.25USD / Case of 960
VistaLab VistaRak™ Pipette Tips, Sterile, RNase/DNase Free, 250 µL; 192 Tips x 5 Racks EW-06197-77 Mfr # 4060-2132
COMPARE
250 Yes Small; RNase/DNase Free
$115.00USD / Case of 960
VistaLab VistaRak™ Pipette Tips, Sterile, RNase/DNase Free, 1250 µL; 192 Tips x 5 Racks EW-06197-78 Mfr # 4060-3132
COMPARE
1250 Yes Large; RNase/DNase Free
$134.75USD / Case of 960
VistaLab VistaRak™ Pipette Tips, Sterile, Filtered, RNase/DNase Free, 25 µL; 192 Tips x 5 Racks EW-06197-79 Mfr # 4060-1332
COMPARE
25 Yes Micro; RNase/DNase Free & Filtered
$172.00USD / Case of 960
VistaLab VistaRak™ Pipette Tips, Sterile, Filtered, RNase/DNase Free, 25 µL; 192 Tips x 5 Racks EW-06197-80 Mfr # 4060-1333
COMPARE
25 Yes Micro; RNase/DNase Free & Filtered
$242.00USD / Case of 960
VistaLab VistaRak™ Pipette Tips, Sterile, Filtered, RNase/DNase Free, 250 µL; 192 Tips x 5 Racks EW-06197-81 Mfr # 4060-2332
COMPARE
250 Yes Small; RNase/DNase Free & Filtered
$166.00USD / Case of 960
VistaLab VistaRak™ Pipette Tips, Sterile, Filtered, RNase/DNase Free, 1250 µL; 192 Tips x 5 Racks EW-06197-82 Mfr # 4060-3332
COMPARE
1250 Yes Large; RNase/DNase Free & Filtered
$194.25USD / Case of 960
Showing 1- 10 of 10