VistaLab™ VistaStak™ Non-Sterile Pipette Tips
VistaStak™ style reduces lab bench clutter by combining multiple layers into one rack
  • Tips made of 100% pure virgin polypropylene
  • Environmentally friendly design uses less plastic
  • Save space—192 tips per layer with a footprint of just 4.5” x 6”
  • Each layer is securely locked to a lower layer for exceptional stability
VistaStak™ polypropylene (PP) pipette tips come in a stacked rack layout. Each layer of this unique design features a hinged lock to the next lower layer with a reusable hinged top for each later as it’s uncovered. 250 and 1250 µL tips are compatible with VistaLab™ Ovation and MLA model pipettors. 25 µL tips are only compatible with VistaLab™ Ovation model pipettors.
$60.30 - $85.50USD / Pkg of 576
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (µL) Package Of Sterile
Availability Pricing
VistaLab VistaStak™ Pipette Tips, Non-Sterile, 25 µL; 192 Tips x 3 Layers EW-06197-73 Mfr # 4060-9024
COMPARE
25 3 Layer Rack; 576/Pk No
$60.30USD / Pkg of 576
VistaLab VistaStak™ Pipette Tips, Non-Sterile, 250 µL; 192 Tips x 5 Layers EW-06197-74 Mfr # 4060-9025
COMPARE
250 5 Layer Rack; 960/Pk No
$85.50USD / Pkg of 960
VistaLab VistaStak™ Pipette Tips, Non-Sterile, 1250 µL; 192 Tips x 3 Layers EW-06197-75 Mfr # 4060-9026
COMPARE
1250 3 Layer Rack; 576/Pk No
$74.10USD / Pkg of 576