Waring Two-speed Laboratory Blenders
Two-speed blenders feature push-button speed control for 18,000 and 22,000 rpm operation. All models include a container, lid, and stainless steel (SS) blades. The 120 VAC models are UL-listed and have a 6-ft cord with U.S. standard plug; 240 VAC models are CE-marked and have a 6-ft cord only.
$1,065.00 - $1,260.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (L) Container Material Max RPM
Availability Pricing
Waring 7011S 2-Speed Blender, 1L, Stainless Steel Container, 120 VAC -- Standard Motor EW-04241-10 Mfr # 7011S
COMPARE
1 Stainless Steel 22000
$1,065.00USD / Each
Waring 7011HG 2-Speed Blender, 1L, Glass, 120 VAC -- Heavy-Duty Motor EW-04241-20 Mfr # 7011HG
COMPARE
1 Glass 22000
$1,170.00USD / Each
Waring 7011HS 2-Speed Heavy-Duty Lab Blender with 1 L SS Container; 120 VAC EW-04241-22 Mfr # 7011HS
COMPARE
1 Stainless Steel 22000
$1,260.00USD / Each
Waring 7011G 2-Speed Blender, 1L, Glass Container, 120 VAC -- Standard Motor EW-04242-10 Mfr # 7011G
COMPARE
1 Glass 22000
$1,090.00USD / Each