Water-Jet Aspirator Pump
Produce pulseless vacuums up to 29.5"
  • Built-in circulating pump and water tank makethese units portable and eliminate water waste
  • Simple to operate—just fill the tank and turn on the pump
  • Circulating pump continuously forces water across a set of venturies—operation is quiet,
  • clean and efficient.
Pump features two metal venturi aspiration jets to create the vacuum with a built-in check valve to prevent back-flow of water into the system. Tank features a drain port and spigot for easy water changes. Order the optional vacuum gauge and regulator to monitor and control vacuum level.
$1,685.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC)
Availability Pricing
COMPARE
115
$1,685.00USD / Each
COMPARE
230
$1,685.00USD / Each