Watson-Marlow 120S Variable-Speed Pump System
Compact pump systems deliver accuracy and repeatability to optimize your fluid handling process
  • Small, stackable pumps require minimal benchtop space
  • Simple operation—requires minimal key presses
  • Control and view speed on digital LCD
  • Keypad lock prevents setting accidental changes
  • Smooth IP31-rated housing for easy wipe down
  • Auto start for automatic power recovery
Easily maintain fluid integrity using peristaltic pumps. Only the bore of the tubing contacts the fluid, eliminating the risk of the pump contaminating the fluid or the fluid contaminating the pump. Peristaltic pumps are great for use with most liquids, including viscous, shear-sensitive, corrosive, and abrasive fluids.
Manual control drives include a single-channel pump head for flow rates up to 170 mL/min*—select from 2-roller, 4-roller, or 4-roller flip-top models. Easily adjust speed, start/stop, and direction from the keypad. Adjust speed in 1 rpm increments using the up/down arrows. Press the MAX key for rapid priming; AUTO START for automatic power recovery. Use the keypad lock feature to prevent any unwanted changes to pump settings. A smooth, IP31-rated housing encases a maintenance-free, brushless motor for quick and easy cleaning. Contact us for free technical application support on pump and tubing selection.
$2,700.00 - $3,200.00USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (mL/min) Min Flow Rate (mL/min) Pump Head
Availability Pricing
Watson-Marlow 120S/D1 Variable-Speed Pump System, Manual Control, 1 to 200 rpm, Single-Channel, 4-Roller; 90 to 264 VAC EW-50130-00 Mfr # 010.6131.010
COMPARE
120 0.001 400D1 four-roller
$2,880.00USD / Each
Watson-Marlow 120S/DV Variable-Speed Pump System, Manual Control, 1 to 200 rpm, Flip-Top Head, Single-Channel, 4-Roller; 90 to 264 VAC EW-50130-01 Mfr # 010.6131.DA0
COMPARE
170 0.02 114DV flip-top four-roller
$2,765.00USD / Each
Watson-Marlow 120S/R Variable-Speed Pump System, Manual Control, 1 to 32 rpm, Single-Channel, 2-Roller; 90 to 264 VAC EW-50130-04 Mfr # 010.6131.R00
COMPARE
54 0.003 102R two-roller
$2,765.00USD / Each
Watson-Marlow 120S/DM2 Variable-Speed Pump System, Manual Control, 1 to 200 rpm, Two-Channel, 4-Roller; 90 to 264 VAC EW-50141-98 Mfr # 010.6131.M20
COMPARE
36 0.0001 400DM2 four-roller
$2,700.00USD / Each
Watson-Marlow 120S/DM3 Variable-Speed Pump System, Manual Control, 1 to 200 rpm, Three-Channel, 4-Roller; 90 to 264 VAC EW-50141-99 Mfr # 010.6131.M30
COMPARE
36 0.0001 400DM3 four-roller
$3,200.00USD / Each