Watson-Marlow 530Du/R2 IP31 Pump System with RS-232 Digital Control
Precision control with RS-232 remote operation capability
  • Dual functionality: digital control and manual operation
  • Flow rates from 0.004 mL/min to 3.5 L/min
  • 2200:1 speed control range from 0.1 to 220 rpm in precise 0.1 rpm increments
  • Auto-restart, leak detection, and configurable logic inputs
  • MemoDose mode allows repeat-dosing of a precise volume of fluid
  • Can operate MemoDose remotely using foot/hand-switch or logic input
  • Full calibration with flow rate and speed display options
  • Secure 3-level PIN lock for enhanced access control
  • IP31 (NEMA 2) cased for protection
The 530Du pump system offers advanced digital and manual control options, ensuring reliable and flexible fluid management. Designed for precision and efficiency, it’s perfect for applications requiring seamless operation and robust security. The 520R2 pumphead features a twin spring roller for use with 2.4 mm wall thickness tubing – tube materials commonly used include Pumpsil® and Bioprene®.
Remote control options include a configurable start/stop, leak detector, and pressure switch input via contact closure, 5 V TTL, or 24 V industrial logic. Direction change and auto/manual toggle input can be made via contact closure, 5 V TTL, or 24 V industrial logic Analog speed control options include fully configurable inputs (0-10 V or 4-20 mA), outputs (0-10 V or 4-20 mA), keypad/analog input scaling, and tacho frequency output (0-991 Hz).
$11,970.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Control Type Max Flow Rate (mL/min) Description
Availability Pricing
Watson-Marlow 530Du/R2 IP31 Pump System, RS-232 Digital Control, US Plug, for 2.4 mm Wall Thickness Tubing EW-50140-01 Mfr # 150.CP51.HU9
COMPARE
Manual, analog, RS-232 3500 IP31 Pump System, RS-232 Digital Control, US Plug, for 2.4 mm Wall Thickness Tubing
$11,970.00USD / Each