Watson-Marlow 530S/R2 IP31 Pump System
Simplify control with the 530S/R pump system
  • Intuitive keypad with color display for easy operation
  • Flow rates from 0.004 mL/min to 3.5 L/min
  • 2200:1 speed control range from 0.1 to 220 rpm in precise 0.1 rpm increments
  • Secure 3-level PIN lock for controlled access
  • Auto-restart and leak detection for uninterrupted performance
  • IP31 (NEMA 2) cased for protection
The 530S pump drive combines precision and ease of use, featuring advanced manual controls for reliable fluid management. The 520R2 pumphead features a twin spring roller for use with 2.4 mm wall thickness tubing – tube materials commonly used include Pumpsil® and Bioprene®. Ideal for environments demanding accuracy and security, it ensures seamless operation with minimal effort.
$7,260.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Control Type Max Flow Rate (mL/min) Description
Availability Pricing
Watson-Marlow 530S/R2 IP31 Pump System, US Plug, for 2.4 mm Wall Thickness Tubing EW-50140-00 Mfr # 150.CP31.HU8
COMPARE
Manual 3500 IP31 Pump System, US Plug, for 2.4 mm Wall Thickness Tubing
$7,260.00USD / Each