Watson-Marlow Fixed-Speed Pump Systems
Simple-to-use fixed-speed pump drive feature superior speed control for accuracy and repeatable flow
  • Small, stackable pumps require minimal benchtop space
  • Simple operation—requires minimal key presses
  • Smooth IP31-rated housing for easy wipe down
Easily maintain fluid integrity using peristaltic pumps. Only the bore of the tubing contacts the fluid, eliminating the risk of the pump contaminating the fluid or the fluid contaminating the pump. Peristaltic pumps are great for use with most liquids, including viscous, shear-sensitive, corrosive, and abrasive fluids.
Achieve repeatable flow every time with these fixed-speed pumps featuring excellent speed accuracy. Simply press green START key to start the pump and press the red STOP key to stop the pump. Compact pumps are stackable, saving you valuable benchtop space. A smooth, IP31-rated housing encases a maintenance-free, brushless motor for quick and easy cleaning.
$1,920.00USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (mL/min) Max RPM Pump Head
Availability Pricing
Watson-Marlow 120F/R Fixed-Speed Pump System, 10 rpm, Single-Channel, 2-Roller Pump Head; 90 to 264V EW-50141-90 Mfr # 010.6111.R00
COMPARE
17 10 102R two-roller
$1,920.00USD / Each
Watson-Marlow 120/F/DV Fixed-Speed Pump System, 10 rpm, Single-Channel, 4-Roller Pump Head; 90 to 264V EW-50141-91 Mfr # 010.6111.DA0
COMPARE
8.5 10 114DV flip-top four-roller
$1,920.00USD / Each
Watson-Marlow 120F/R Fixed-Speed Pump System, 17 rpm, Single-Channel, 2-Roller Pump Head; 90 to 264V EW-50141-92 Mfr # 010.6112.R00
COMPARE
29 17 102R two-roller
$1,920.00USD / Each
Watson-Marlow 120F/DV Fixed-Speed Pump System, 17 rpm, Single-Channel, 4-Roller Pump Head; 90 to 264V EW-50141-93 Mfr # 010.6112.DA0
COMPARE
14 17 114DV flip-top four-roller
$1,920.00USD / Each
Watson-Marlow 120F/R Fixed-Speed Pump System, 31 rpm, Single-Channel, 2-Roller Pump Head; 90 to 264V EW-50141-94 Mfr # 010.6113.R00
COMPARE
52 31 102R two-roller
$1,920.00USD / Each
Watson-Marlow 120F/DV Fixed-Speed Pump System, 31 rpm, Single-Channel, 4-Roller Pump Head; 90 to 264V EW-50141-95 Mfr # 010.6113.DA0
COMPARE
26 31 114DV flip-top four-roller
$1,920.00USD / Each
Watson-Marlow 120F/DV Fixed-Speed Pump System, 52 rpm, Single-Channel, 4-Roller Pump Head; 90 to 264V EW-50141-96 Mfr # 010.6114.DA0
COMPARE
44 52 114DV flip-top four-roller
$1,920.00USD / Each
Watson-Marlow 120F/DV Fixed-Speed Pump System, 220 rpm, Single-Channel, 4-Roller Pump Head; 90 to 264V EW-50141-97 Mfr # 010.6115.DA0
COMPARE
190 220 114DV flip-top four-roller
$1,920.00USD / Each
Showing 1- 8 of 8