Watson-Marlow Foot and Hand Switches
Enhance your Watson-Marlow peristaltic pump operations with specialized control accessories, designed for precision and ease of use in demanding laboratory and industrial environments
  • Equipped with a 25-pin D-connector, connect directly to models 323Du and 520Du for seamless integration
  • Use the footswitch for hands-free operation that allows for multitasking and maintains sterile conditions
  • Use the handswitch for precise manual control with a comfortable ergonomic design
Optimize your fluid handling processes with these essential accessories, ensuring enhanced control and efficiency in your applications. Both the hand and footswitch will operate the MemoDose function on Watson-Marlow pumps 323Du and 520Du.
$229.00 - $679.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Watson-Marlow 520Af Footswitch EW-50135-06 Mfr # 059.3002.000
COMPARE
Footswitch
$679.00USD / Each
Watson-Marlow 520Ah Handswitch EW-50135-07 Mfr # 059.3022.000
COMPARE
Handswitch
$229.00USD / Each