Watson-Marlow Peroxide-Cured Silicone 3-Stop Pump Tubing
Use with any Watson-Marlow multichannel cartridges
  • Stops ensure a perfect fit every time
  • Color-coded stops for easy identification
  • Exhibits good heat, cold, ozone, UV, and abrasion resistance
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
Peroxide-cured silicone tubing has hardness shore of 60A ±5 and elongation at break ranging from 440 to 530% to provide a long shelf life and enhanced mechanical strength—ideal for use in peristatic pumps. Stops on tubing are color-coded according to tubing ID and ensure a perfect fit on your pump head. Three stops on the tubing offer two pump segments, doubling the life of the tubing. Silicone tubing is available with tubing ID from 0.63 to 2.79 mm.
$315.00USD / Pkg of 6
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Coded Stops Tubing ID (mm)
Availability Pricing
Watson-Marlow Silicone 3-Stop Pump Tubing, 0.63 mm, Orange/White; 6/Pk EW-50130-71 Mfr # 983.0063.000
COMPARE
Orange/White 0.63
$315.00USD / Pkg of 6
Watson-Marlow Silicone 3-Stop Pump Tubing, 0.76 mm, Black/Black; 6/Pk EW-50130-72 Mfr # 983.0076.000
COMPARE
Black/Black 0.76
$315.00USD / Pkg of 6
Watson-Marlow Silicone 3-Stop Pump Tubing, 0.88 mm, Orange/Orange; 6/Pk EW-50130-73 Mfr # 983.0088.000
COMPARE
Orange/Orange 0.88
$315.00USD / Pkg of 6
Watson-Marlow Silicone 3-Stop Pump Tubing, 1.02 mm, White/White; 6/Pk EW-50130-74 Mfr # 983.0102.000
COMPARE
White/White 1.02
$315.00USD / Pkg of 6
Watson-Marlow Silicone 3-Stop Pump Tubing, 1.14 mm, Red/Red; 6/Pk EW-50130-75 Mfr # 983.0114.000
COMPARE
Red/Red 1.14
$315.00USD / Pkg of 6
Watson-Marlow Silicone 3-Stop Pump Tubing, 1.29 mm, Grey/Grey; 6/Pk EW-50130-76 Mfr # 983.0129.000
COMPARE
Grey/Grey 1.29
$315.00USD / Pkg of 6
Watson-Marlow Silicone 3-Stop Pump Tubing, 1.42 mm, Yellow/Yellow; 6/Pk EW-50130-77 Mfr # 983.0142.000
COMPARE
Yellow/Yellow 1.42
$315.00USD / Pkg of 6
Watson-Marlow Silicone 3-Stop Pump Tubing, 1.52 mm, Yellow/Blue; 6/Pk EW-50130-78 Mfr # 983.0152.000
COMPARE
Yellow/Blue 1.52
$315.00USD / Pkg of 6
Watson-Marlow Silicone 3-Stop Pump Tubing, 1.65 mm, Blue/Blue; 6/Pk EW-50130-79 Mfr # 983.0165.000
COMPARE
Blue/Blue 1.65
$315.00USD / Pkg of 6
Watson-Marlow Silicone 3-Stop Pump Tubing, 1.85 mm, Green/Green; 6/Pk EW-50130-80 Mfr # 983.0185.000
COMPARE
Green/Green 1.85
$315.00USD / Pkg of 6
Watson-Marlow Silicone 3-Stop Pump Tubing, 2.05 mm, Purple/Purple; 6/Pk EW-50130-81 Mfr # 983.0205.000
COMPARE
Purple/Purple 2.05
$315.00USD / Pkg of 6
Watson-Marlow Silicone 3-Stop Pump Tubing, 2.29 mm, Purple/Black; 6/Pk EW-50130-82 Mfr # 983.0229.000
COMPARE
Purple/Black 2.29
$315.00USD / Pkg of 6
Watson-Marlow Silicone 3-Stop Pump Tubing, 2.54 mm, Purple/Orange; 6/Pk EW-50130-83 Mfr # 983.0254.000
COMPARE
Purple/Orange 2.54
$315.00USD / Pkg of 6
Watson-Marlow Silicone 3-Stop Pump Tubing, 2.79 mm, Purple/White; 6/Pk EW-50130-84 Mfr # 983.0279.000
COMPARE
Purple/White 2.79
$315.00USD / Pkg of 6
Showing 1- 14 of 14