Welch High Vacuum Pump
Handles harshest absorption chiller servicing. The DuoSeal belt drive high vacuum pumps are known worldwide for ruggedness and dependability. These pumps have large oil reservoirs for their pumping capacity to dilute contaminates coupled with low pump RPM to reduce friction and wear.
FEATURES
  • Low pump RPM
  • High contamination tolerance
  • Seals and gaskets for newest refrigerants
$4,895.00 - $9,175.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Free Air Capacity (CFM)
Availability Pricing
Welch 1376B-46 Hi Vacuum Pump 10.6CFM 115v 60Hz 1Ph EW-68479-29 Mfr # 1376B-46
COMPARE
10.6
$6,750.00USD / Each
Welch 1397B-46 Hi Vacuum Pump 17.7CFM 115v 60Hz 1Ph EW-68479-30 Mfr # 1397B-46
COMPARE
17.7
$9,175.00USD / Each
Welch 1402B-46 Hi Vacuum Pump 4.6CFM 115v 60Hz EW-68479-31 Mfr # 1402B-46
COMPARE
4.6
$4,895.00USD / Each