Liquid Level Control Selection Guide
Liquid Level Control Selection Guide
Choose the correct liquid level option for your application
Single-Point Control    Dual-Point Control     Continuous-Level Control     Leak Detection
Single-Point Control
Single-point control
  • Common application: Keep tank from overflowing or running dry.
  • Compatible sensor types: Float, capacitance, optical, proximity, tuning fork, ultrasonic
  • How it works: Each time the liquid reaches a critical level, the sensor turns on a pump or opens a valve to prevent the tank from overflowing/running dry.
Dual-Point Control
  • Common application: Keep tank filled between two critical points.
  • Compatible sensor types: Same as for single-point control (above).
  • How it works: Install sensors at two critical points. If liquid falls below the lower sensor, the detector activates a pump until liquid reaches the upper sensor.
Continuous-Level Control
Continuous-level control
  • Common application: Control level at all points and times, possibly activating a pump, valve, or alarm.
  • Compatible sensor types: Symprobe™, Cricket™, ultrasonic, radar wave
  • How it works: Continuous-level sensors have a continuous analog output that is proportional to the level at all times. Level may be recorded with an external device.
Leak Detection
Leak detection
  • Common application: Detect leak in a pipe, possibly activating a pump, valve, or alarm.
  • Compatible sensor types: Optical, proximity
  • How it works: Install one or more sensors around a container or pipe. If the sensor detects liquid, it activates alarms or pump systems.