Bel-Art Autoclavable HDPE Biohazard Bags, 20 to 30 Gallon, 31 x 38"; 200/CS

Representative image only.

Bel-Art Autoclavable HDPE Biohazard Bags, 20 to 30 Gallon, 31 x 38"; 200/CS

Bel-Art | Mfr: F13164-3138| Item: EW-06494-06

Hold more—up to 50 gallons

$464.50USD / Case of 200

$464.50USD / Case of 200
Product Description

Key Features

  • No misinterpretations—procedures printed in four languages
  • Know when it's autoclaved—indicator
  • patch darkens for verification
  • Autoclavable at 250°F (121°C).

More About this Item

Bright orange-red, 1.25 mil, high-molecular weight HDPE bags are printed with the biohazard warning symbol and precautionary procedures in four languages. The sterilization indicator patch darkens when autoclaved.

Specifications
  • MaterialHDPE
  • Capacity (Gal)30
  • ColorOrange and Red
  • Length (in)38
  • Width (in)31
  • Length (cm)96.52
  • Width (cm)78.74
  • Thickness (mil)1
  • Description20 to 30 Gallon Autoclavable HDPE Biohazard Bags, 31 x 38"; 200/cs
Reviews
Q&A