Bel-Art Biohazard Disposal Bags, HDPE

Hold more—up to 50 gallons
  • No misinterpretations—procedures printed in four languages
  • Know when it's autoclaved—indicator
  • patch darkens for verification
  • Autoclavable at 250°F (121°C).
Bright orange-red, 1.25 mil, high-molecular weight HDPE bags are printed with the biohazard warning symbol and precautionary procedures in four languages. The sterilization indicator patch darkens when autoclaved.
$136.50 - $464.50USD / Case of 200

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (Gal) Material Thickness (mil)
Availability Pricing
Bel-Art Autoclavable HDPE Biohazard Bags, 2 to 4 Gallon, 14 x 19"; 200/CS EW-06494-00 Mfr # F13164-1419
COMPARE
4 HDPE 1
$136.50USD / Case of 200
Bel-Art Autoclavable HDPE Biohazard Bags, 6 to 9 Gallon, 19 x 23"; 200/CS EW-06494-02 Mfr # F13164-1923
COMPARE
9 HDPE 1
$199.50USD / Case of 200
Bel-Art Autoclavable HDPE Biohazard Bags, 20 to 30 Gallon, 31 x 38"; 200/CS EW-06494-06 Mfr # F13164-3138
COMPARE
30 HDPE 1
$464.50USD / Case of 200
Bel-Art Autoclavable HDPE Biohazard Bags, 40 to 50 Gallon, 38 x 48"; 100/CS EW-06494-08 Mfr # F13164-3848
COMPARE
50 HDPE 1
$360.50USD / Case of 100
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat