Bel-Art Autoclavable HDPE Biohazard Bags, 6 to 9 Gallon, 19 x 23"; 200/CS

Representative image only.

Bel-Art Autoclavable HDPE Biohazard Bags, 6 to 9 Gallon, 19 x 23"; 200/CS

Bel-Art | Mfr: F13164-1923| Item: EW-06494-02

Hold more—up to 50 gallons

$199.50USD / Case of 200

$199.50USD / Case of 200
Product Description

Key Features

  • No misinterpretations—procedures printed in four languages
  • Know when it's autoclaved—indicator
  • patch darkens for verification
  • Autoclavable at 250°F (121°C).

More About this Item

Bright orange-red, 1.25 mil, high-molecular weight HDPE bags are printed with the biohazard warning symbol and precautionary procedures in four languages. The sterilization indicator patch darkens when autoclaved.

Specifications
  • ColorOrange and Red
  • Capacity (Gal)9
  • MaterialHDPE
  • Length (in)23
  • Width (in)19
  • Length (cm)58.42
  • Width (cm)48.26
  • Thickness (mil)1
  • Description6 to 9 Gallon Autoclavable HDPE Biohazard Bags, 19 x 23"; 200/cs
Reviews
Q&A