Bel-Art Autoclavable HDPE Biohazard Bags, 40 to 50 Gallon, 38 x 48"; 100/CS

Representative image only.

Bel-Art Autoclavable HDPE Biohazard Bags, 40 to 50 Gallon, 38 x 48"; 100/CS

Bel-Art | Mfr: F13164-3848| Item: EW-06494-08

Hold more—up to 50 gallons

$360.50USD / Case of 100

$360.50USD / Case of 100
Product Description

Key Features

  • No misinterpretations—procedures printed in four languages
  • Know when it's autoclaved—indicator
  • patch darkens for verification
  • Autoclavable at 250°F (121°C).

More About this Item

Bright orange-red, 1.25 mil, high-molecular weight HDPE bags are printed with the biohazard warning symbol and precautionary procedures in four languages. The sterilization indicator patch darkens when autoclaved.

Specifications
  • ColorOrange and Red
  • MaterialHDPE
  • Capacity (Gal)50
  • Length (in)48
  • Width (in)38
  • Length (cm)121.92
  • Width (cm)96.52
  • Thickness (mil)1
  • Description40 to 50 Gallon Autoclavable HDPE Biohazard Bags, 38 x 48"; 100/cs
Reviews
Q&A