Celite 545 Filter Aid, Powder, 50 lb

Representative image only.

Celite 545 Filter Aid, Powder, 50 lb

Celite | Mfr: C21250LB| Item: EW-88068-93

Reagents and solutions to meet your lab and research needs

$1,851.00USD / Each

$1,851.00USD / Each
Product Description

Key Features

  • Quality and consistency your lab can count on

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Storage Code Gray
Specifications
  • Empirical FormulaCNa2O3
  • SynonymsCalcinated diatomaceous earth, Diatomite,Diatomaceous Earth, Kieselguhr
  • FormPowder
  • Weight (g)22680
  • Formula Weight105.988
  • PackagingFiber Drum
  • DOT InformationNot regulated by DOT
  • CAS Number68855-54-9
  • Description545 Filter Aid, Powder, 50 lb
Documents & Certificates
Reviews
Q&A