Celite™ 545 Filter Aid

Reagents and solutions to meet your lab and research needs
  • Quality and consistency your lab can count on
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$132.75 - $1,851.00USD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
CAS Number Weight (g) Description
Availability Pricing
Celite 545 Filter Aid, Powder, 25 lb EW-88068-91 Mfr # C21225LB
COMPARE
68855-54-9 11340 545 Filter Aid, Powder, 25 lb
$1,221.00USD / Each
Celite 545 Filter Aid, Powder, 50 lb EW-88068-93 Mfr # C21250LB
COMPARE
68855-54-9 22680 545 Filter Aid, Powder, 50 lb
$1,851.00USD / Each
Celite 545 Filter Aid, Powder, 2.5 kg EW-88320-32 Mfr # AC349670025
COMPARE
68855-54-9 2500 545 Filter Aid, Powder, 2.5 kg
$268.00USD / Each
Celite 545 Filter Aid, Powder, 500 g EW-88320-33 Mfr # AC349675000
COMPARE
68855-54-9 500 545 Filter Aid, Powder, 500 g
$132.75USD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat