Cole-Parmer Acetic Acid, Glacial, ACS Grade, Clear Glass, 500 mL

Representative image only.

Cole-Parmer Acetic Acid, Glacial, ACS Grade, Clear Glass, 500 mL

Cole-Parmer | Item: EW-88401-65

Reagents and solutions to meet your lab and research needs

$101.50USD / Each

$101.50USD / Each

Buy It Together

Current Item

Cole-Parmer Acetic Acid, Glacial, ACS Grade, Clear Glass, 500 mL
Cole-Parmer Acetic Acid, Glacial, ACS Grade, Clear Glass, 500 mL
8840165

$101.50USD / Each
+ =
Total Price 195.60 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Quality and consistency your lab can count on

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeACS Reagent
  • Concentration1
  • Empirical FormulaCH3COOH
  • ClassReagent Acid
  • FormLiquid
  • Volume (mL)500
  • Formula Weight60.05
  • PackagingClear Glass
  • DOT InformationAcetic acid, glacial; Class 8 (3), Packing Group II
  • Storage SymbolRed
  • Hazard SymbolCorrosive
  • UN NumberUN2789
  • CAS Number64-19-7
  • Shelf Life (days)60
  • DescriptionAcetic Acid, Glacial, ACS Grade, Clear Glass, 500 mL
Reviews
Q&A