Corning 9989 9989 Fiber Glass Wool, 8 um porosity, 12 lb/pk
Representative image only.
Corning 9989 9989 Fiber Glass Wool, 8 um porosity, 12 lb/pk
Corning | Mfr: 3950| Item: EW-34552-21
Suitable as a filtering medium
$1,795.00USD / Case of 1
BUY MORE & SAVE
  • 1-4$1,795.00 USD / Case of 1
  • 5-9$1,615.50 USD / Case of 1
  • 10+$1,525.75 USD / Case of 1
$1,795.00USD / Case of 1
BUY MORE & SAVE
  • 1-4$1,795.00 USD / Case of 1
  • 5-9$1,615.50 USD / Case of 1
  • 10+$1,525.75 USD / Case of 1
Product Description
Key Features
  • Strong,yet soft and pliable
  • Contains minimal heavy metals
  • Easy to use
  • Available 12 lb/pack
More About this Item
Pliable glass wool is manufactured from code no. 9989 glass and contains minimal heavy metals — suitable for use as a filtering medium. Fine glass fibers have diameters controlled to approximately 8 µm. Fiber glass wool is supplied in the form of roping approximately 2" dia x 22 ft long per 1-lb pack.
Specifications
  • MaterialFiberglass
  • Description9989 Fiber Glass Wool, 8 um porosity, 12 lb/pk