Pyrex Fiber Glass Wool
Suitable as a filtering medium
  • Strong,yet soft and pliable
  • Contains minimal heavy metals
  • Easy to use
Pliable glass wool is manufactured from code no. 9989 glass and contains minimal heavy metals — suitable for use as a filtering medium. Fine glass fibers have diameters controlled to approximately 8 µm. Fiber glass wool is supplied in the form of roping approximately 2" dia x 22 ft long per 1-lb pack.
$204.00 - $1,795.00USD / Case of 1
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Material Description
Availability Pricing
Corning 9989 9989 Fiber Glass Wool, 8 um porosity, 2/pk
Buy More Save More
EW-34552-01 Mfr # 3950
COMPARE
Fiberglass 9989 Fiber Glass Wool, 8 um porosity, 2/pk
$204.00USD / Pkg of 1
Corning 9989 9989 Fiber Glass Wool, 8 um porosity, 12 lb/pk
Buy More Save More
EW-34552-21 Mfr # 3950
COMPARE
Fiberglass 9989 Fiber Glass Wool, 8 um porosity, 12 lb/pk
$1,795.00USD / Case of 1