Environmental Express 5% Cesium Ionization Buffer for FAAS in 1% HNO3; 500 mL

Representative image only.

Environmental Express 5% Cesium Ionization Buffer for FAAS in 1% HNO3; 500 mL

Environmental Express | Item: EW-97015-63

Use buffer to increase the free electron population in flame emission or absorption

$366.00USD / Each

$366.00USD / Each
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Designed for use with flame atomic absorption spectroscopy (FAAS)
  • Buffers help suppress ionization interference effects
  • Recommended by manufacturers of ICP and AAS instrumentation
  • 5% cesium ionization buffer in 1% HNO3 matrix
  • 500 mL

More About this Item

Can’t find the standards you are looking for? HPS can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 843-767-7900.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous material shipping fees may apply.

Specifications
  • Concentration5%
  • Matrix1% HNO3
  • Element/IonCesium (Cs)
  • FormLiquid
  • Volume (mL)500
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s. (Nitric acid); Class 8, Packing Group III
  • Hazard SymbolIrritant
  • UN NumberUN3264
  • Shelf Life (months)24
  • Description5% Cesium Ionization Buffer for FAAS in 1% HNO3; 500 mL
Documents & Certificates
Reviews
Q&A