Environmental Express® 5% Cesium Ionization Buffers for FAAS in 1% HNO3
Use buffer to increase the free electron population in flame emission or absorption
  • Designed for use with flame atomic absorption spectroscopy (FAAS)
  • Buffers help suppress ionization interference effects
  • Recommended by manufacturers of ICP and AAS instrumentation
Can’t find the standards you are looking for? HPS can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 843-767-7900.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$218.00 - $366.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Concentration Form Description
Availability Pricing
COMPARE
5% Liquid 5% Cesium Ionization Buffer for FAAS in 1% HNO3; 500 mL
$366.00USD / Each
COMPARE
5% Liquid 5% Cesium Ionization Buffer for FAAS in 1% HNO3; 250 mL
$218.00USD / Each