Fisher Chemical 1-Butanol, Certified ACS Grade, 4 L

Representative image only.

Fisher Chemical 1-Butanol, Certified ACS Grade, 4 L

Fisher Chemical | Mfr: A3994| Item: EW-88003-90

Quality and consistency your lab can count on

$407.00USD / Each

$407.00USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Assay — ≥99.4%
  • Carbonyl Compounds (as Butyraldehyde) — ≤0.01%
  • Color (APHA) — ≤10
  • Titratable Acid — ≤0.0008mEq/g
  • Residue after Evaporation — ≤0.005%
  • Water — ≤0.1%
  • Fluorescent Impurities (as Quinine Sulfate) — To pass test (≤0.02μg/mL)
  • Butyl Ether (by GC) — ≤0.2%
  • Boiling Range (1 drop to dryness) — ≤1.5°C
  • Boiling Point — 117.7°C ± 0.1°C

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

HarmfulStorage Code Red
Specifications
  • Purity GradeCertified
  • Empirical FormulaCH3CH2CH2CH2OH
  • ClassReagent Solvents
  • ChemistrySolvents
  • Synonymsn-Propyl Carbinol, n-Butyl Alcohol
  • FormLiquid
  • Volume (mL)4000
  • Formula Weight74.12
  • PackagingGlass Bottle
  • DOT InformationBUTANOLS; Class 3, Packing Group III
  • Storage SymbolRed
  • Hazard SymbolHarmful
  • CAS Number71-36-3
  • UN NumberUN1120
  • Boiling Point C (° C)117.6
  • Melting Point C (° C)-89
  • Description1-Butanol, Certified ACS Grade, 4 L
Documents & Certificates
Reviews
Q&A