Fisher Chemical™ 1-Butanol

Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$374.75 - $606.00USD / Each

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Purity Grade Volume (mL) Description
Availability Pricing
Fisher Chemical 1-Butanol, HPLC Grade, 1 L EW-88003-82 Mfr # A3831
COMPARE
HPLC 1000 1-Butanol, HPLC Grade, 1 L
$374.75USD / Each
Fisher Chemical 1-Butanol, HPLC Grade, 4 L EW-88003-84 Mfr # A3834
COMPARE
HPLC 4000 1-Butanol, HPLC Grade, 4 L
$606.00USD / Each
Fisher Chemical 1-Butanol, Certified ACS Grade, 4 L EW-88003-90 Mfr # A3994
COMPARE
Certified 4000 1-Butanol, Certified ACS Grade, 4 L
$407.00USD / Each
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat