Fluke 53-2 B 60HZ Single-Input Datalogging Thermocouple Thermometer
Representative image only.
Fluke 53-2 B 60HZ Single-Input Datalogging Thermocouple Thermometer
Fluke | Mfr: FLUKE-53-2 B 60HZ| Item: EW-91500-08
Rugged design with splash- and dust-resistant housing
$573.30USD / Each
$637.00USD / Each
for an additional
$212.25USD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$573.30USD / Each
$637.00USD / Each
Buy It Together
Current Item
Fluke 53-2 B 60HZ Single-Input Datalogging Thermocouple Thermometer
Fluke 53-2 B 60HZ Single-Input Datalogging Thermocouple Thermometer
9150008
$573.30USD / Each
$637.00USD / Each
+ + + + =
Total Price 1185.15 ADD 5 ITEMS TO CART
Product Description
Includes
one type K bead thermocouple per input channel and protective holster.
Key Features
  • Readout in °F, °C, or Kelvin (K)
  • Meters are ruggedly designed with a splash- and dust-resistant housing.
  • Sleep mode increases battery life
  • 3 year warranty
More About this Item
Dual backlit display provides readout of channel temperature, differential temperature (dual-input models), and minimum/maximum/average readings with time stamp.
Store up to 500 points with adjustable recording interval; recall on display or download to your computer through the IR data port.
Available for sale in the United States and Canada only.
Canadian Standards Association3 Year Warranty
Specifications
  • Meter Measurement TypeThermocouple
  • Number Of Probe Channels1
  • Types Of Probes AcceptedType E, Type J, Type K, Type N, Type R, Type S, Type T
  • Min Temperature (° F)-418
  • Max Temperature (° F)3216
  • Min Temperature (° C)-250
  • Max Temperature (° C)1767
  • AccuracyType E, J, K, N, T: ±0.05% of reading +0.5°F (+0.3°C)
  • Resolution0.1°; 1° above 1000°
  • Data LoggingYes
  • Memory Capacity500 data points
  • Height (in)6 7/8
  • Battery Life1000 hours
  • Battery3 AA (included)
  • DescriptionSingle-Input Datalogging Thermocouple Thermometer