Hach 2793844 Total Hardness Test Strips, 0 to 425 mg/L; 250 Strips/Pk
Representative image only.
Hach 2793844 Total Hardness Test Strips, 0 to 425 mg/L; 250 Strips/Pk
Hach | Mfr: 2793844| Item: EW-05546-28
Reduce risk by using as a pre-test validation tool
$176.00USD / Each
$176.00USD / Each
Buy It Together
Current Item
Hach 2793844 Total Hardness Test Strips, 0 to 425 mg/L; 250 Strips/Pk
Hach 2793844 Total Hardness Test Strips, 0 to 425 mg/L; 250 Strips/Pk
0554628
$176.00USD / Each
+ + + + =
Total Price 320.30 ADD 5 ITEMS TO CART
Customers Also Bought
Product Description
Includes
test strips and color key for reading results.
Key Features
  • Simplified testing with results in less than one minute
  • Test strips eliminate chemical handling and cleanup
  • Vial label contains the color key for easy readings in lab or field
More About this Item
Test hardness as CaC)3 in the range of 0 to 425 mg/L ( 0 to 25 gpg). 250 individually wrapped test strips included.
ISO9001 Certified Supplier
Specifications
  • TestHardness, calcium carbonate
  • Measurement Points0, 25, 50, 120, 250, 425 ppm
  • Reagent PlatformStrips
  • Tests Per Kit250
  • DescriptionTotal Hardness Test Strips, 0 to 425 mg/L; 250 Strips/Pk