Hach 2793944 Free and Total Chlorine Low Range Test Strips, 250
Representative image only.
Hach 2793944 Free and Total Chlorine Low Range Test Strips, 250
Hach | Mfr: 2793944| Item: EW-05546-20
Reduce risk by using as a pre-test validation tool
$353.00USD / Each
$353.00USD / Each
Buy It Together
Current Item
Hach 2793944 Free and Total Chlorine Low Range Test Strips, 250
Hach 2793944 Free and Total Chlorine Low Range Test Strips, 250
0554620
$353.00USD / Each
+ + + + =
Total Price 497.30 ADD 5 ITEMS TO CART
Customers Also Bought
Product Description
Includes
test strips and color key for reading results.
Key Features
  • Simplified testing with results in less than one minute
  • Test strips eliminate chemical handling and cleanup
  • Vial label contains the color key for easy readings in lab or field
More About this Item
Tests free and total chlorine as Cl2 from 0 to 10 mg/L. 250 individually wrapped test strips included.
ISO9001 Certified Supplier
Specifications
  • TestChlorine, free and total; low range
  • Measurement Points0, 0.5, 1, 2, 4, 10 ppm
  • Reagent PlatformStrips
  • Tests Per Kit250
  • DescriptionFree and Total Chlorine Low Range Test Strips, 250