High Five N192 Nitrile Gloves, Medium; 1000/CS
Representative image only.
High Five N192 Nitrile Gloves, Medium; 1000/CS
High Five | Mfr: N192| Item: EW-86480-56
Fully textured surface for enhanced grip in wet or dry conditions
$192.00USD / Case of 1000
$192.00USD / Case of 1000
Product Description
Key Features
  • Medical examination grade protects hands of healthcare professionals
  • Tested for use with chemotherapy/cytotoxic drugs
  • Consistent, dependable protection
  • Enhanced tactile sensitivity
  • Powder-free to reduce skin irritation
  • Meets acceptable quality level (AQL) of 1.5
More About this Item
Cobalt gloves are made of soft stretchable nitrile formula that provides a superior fit and feel. They feature a palm thickness of 3.9 mil and a finger thickness of 4.3 mil. Fully texture, the nitrile gloves allow you to securely grip tools and materials — wet or dry.
The multipurpose gloves are ideal for use in the agriculture, life sciences, healthcare, dental, utilities and warehousing industries. They comply with US FDA food contact regulations, so the gloves can also be used in food processing applications. In addition, the gloves help prevent allergic reactions among healthcare workers and patients with latex sensitivities.
Standards and certifications include ASTM D6319, ATM D6978, FDA21 CFR 177-2600-US Food Contact Approved, and US QSR/FDA 510(k) Medical Examination Grade. Ambidextrous.
Specifications
  • MaterialNitrile
  • SizeMedium
  • Length (in)9 1/2
  • Length (cm)24
  • ColorBlue
  • GripTextured
  • CuffBeaded
  • SterileNo
  • PowderedNo
  • Palm Thickness (mil)3.9
  • DescriptionNitrile Gloves, Medium, 1000/cs