High Five Cobalt Nitrile Exam Gloves
Fully textured surface for enhanced grip in wet or dry conditions
  • Medical examination grade protects hands of healthcare professionals
  • Tested for use with chemotherapy/cytotoxic drugs
  • Consistent, dependable protection
  • Enhanced tactile sensitivity
  • Powder-free to reduce skin irritation
  • Meets acceptable quality level (AQL) of 1.5
Cobalt gloves are made of soft stretchable nitrile formula that provides a superior fit and feel. They feature a palm thickness of 3.9 mil and a finger thickness of 4.3 mil. Fully texture, the nitrile gloves allow you to securely grip tools and materials — wet or dry.
The multipurpose gloves are ideal for use in the agriculture, life sciences, healthcare, dental, utilities and warehousing industries. They comply with US FDA food contact regulations, so the gloves can also be used in food processing applications. In addition, the gloves help prevent allergic reactions among healthcare workers and patients with latex sensitivities.
Standards and certifications include ASTM D6319, ATM D6978, FDA21 CFR 177-2600-US Food Contact Approved, and US QSR/FDA 510(k) Medical Examination Grade. Ambidextrous.
$192.00 - $229.60USD / Case of 1000
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Palm Thickness (mil) Size
Availability Pricing
High Five N191 Nitrile Gloves, Small; 1000/CS EW-86480-55 Mfr # N191
COMPARE
9 1/2 3.9 Small
$228.50USD / Case of 1000
High Five N192 Nitrile Gloves, Medium; 1000/CS EW-86480-56 Mfr # N192
COMPARE
9 1/2 3.9 Medium
$192.00USD / Case of 1000
High Five N193 Nitrile Gloves, Large; 1000/CS EW-86480-57 Mfr # N193
COMPARE
9 1/2 3.9 Large
$229.60USD / Case of 1000
High Five N194 Nitrile Gloves, X-Large; 1000/CS EW-86480-58 Mfr # N194
COMPARE
9 1/2 3.9 X-Large
$229.60USD / Case of 1000