Honeywell S3240 Uvex Translucent blue frame glasses with clear lenses
Representative image only.
Honeywell S3240 Uvex Translucent blue frame glasses with clear lenses
Honeywell | Mfr: S3240| Item: EW-86325-80
Available for sale in the US only
$22.10USD / Each
BUY MORE & SAVE
  • 1-9$22.10 USD / Each
  • 10+$19.89 USD / Each
$22.10USD / Each
BUY MORE & SAVE
  • 1-9$22.10 USD / Each
  • 10+$19.89 USD / Each
Product Description
Key Features
  • Sleek, stylish design for cool, comfortable protection
  • Lenses exceed, by seven times, the standards set by ANSI for impact velocity
More About this Item
Wraparound lens design provides uncompromised peripheral vision. Internal points of contact serve as a shock absorber to diffuse impact energy away from eyes.
A ventilation channel between the frame and lens allows moisture to escape — preventing fogging. Coating helps to keep your lenses scratch free. Lenses and temple stems are fully adjustable to provide a custom fit. Compliance: ANSI Z87.1-2010, CSA Z94.3, military V0
Specifications
  • Frame ColorBlue
  • Lens ColorClear
  • Lens CoatingUvextreme Anti-fog
  • ComplianceANSI Z87+ (High Impact); CSA Z94.3; military V0
  • DescriptionTranslucent blue frame glasses with clear lenses