Honeywell S700HS Uvex Stealth Safety Goggles, Clear, HydroShield Antifog Replacement Lens; 10/BX
Representative image only.
Honeywell S700HS Uvex Stealth Safety Goggles, Clear, HydroShield Antifog Replacement Lens; 10/BX
Honeywell | Mfr: S700HS| Item: EW-86497-91
Sleek style, comfort and protection all in one
$97.30USD / Box of 10
$97.30USD / Box of 10
Product Description
Key Features
  • Scratch-resistant and ideal for most indoor work HydroShield long-life antifog coating helps keep lenses clear
  • Replacement Lens HydroShield Antifog
More About this Item
Over-the-Glass (OTG) version is designed to fit comfortably over prescription eyewear. The safety goggle lenses are clear and made of polycarbonate. They provide greater than 99.9% protection from harmful UV rays and come in other lens tints, too.
Compliance: ANSI Z87.1-2015 and CSA Z94.3 standards.
Specifications
  • Lens ColorClear
  • Lens CoatingAntifog
  • Lens TypeStandard
  • ComplianceANSI Z87.1-2015 and CSA Z94.3 standards
  • DescriptionSafety Goggles, Clear, HydroShield Antifog Replacement Lens; 10/BX