Uvex™ by Sperian Stealth® Replacement Lenses for Safety Goggles
Sleek style, comfort and protection all in one
  • Scratch-resistant and ideal for most indoor work HydroShield long-life antifog coating helps keep lenses clear
Over-the-Glass (OTG) version is designed to fit comfortably over prescription eyewear. The safety goggle lenses are clear and made of polycarbonate. They provide greater than 99.9% protection from harmful UV rays and come in other lens tints, too.
Compliance: ANSI Z87.1-2015 and CSA Z94.3 standards.
$97.30 - $279.75USD / Box of 10
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Lens Coating Lens Type Description
Availability Pricing
Honeywell S700HS Uvex Stealth Safety Goggles, Clear, HydroShield Antifog Replacement Lens; 10/BX EW-86497-91 Mfr # S700HS
COMPARE
Antifog Standard Safety Goggles, Clear, HydroShield Antifog Replacement Lens; 10/BX
$97.30USD / Box of 10
Honeywell S740HS Uvex Stealth Safety Goggles, Clear, Over-the-Glass Replacement Lens; 10/BX EW-86497-95 Mfr # S740HS
COMPARE
Antifog Over-the-Glass Safety Goggles, Clear, Over-the-Glass Replacement Lens; 10/BX
$279.75USD / Box of 10