ICI Quick Ship Envision Complete Plumbing Assemblies, Left Side, Air, Orange
Representative image only.
ICI Quick Ship Envision Complete Plumbing Assemblies, Left Side, Air, Orange
ICI Quick Ship | Mfr: CPEISV-A-L| Item: EW-78984-38
Plumbing assemblies designed specifically for Envision workstations
$918.00USD / Each
$918.00USD / Each
Buy It Together
Current Item
ICI Quick Ship Envision Complete Plumbing Assemblies, Left Side, Air, Orange
ICI Quick Ship Envision Complete Plumbing Assemblies, Left Side, Air, Orange
7898438
$918.00USD / Each
+ =
Total Price 1205.75 ADD 2 ITEMS TO CART
Product Description
Includes
Control needle valve service turret with shank, service hose, and a male quick connect fitting
Key Features
  • Conveniently color labeled for easy coupling
  • Control gases up to 60 psi
  • Consist of two major components
  • Available for air, vacuum, and nitrogen services
  • Choose between left and right side assemblies
  • 2 year warranty
More About this Item
Plumbing assemblies for the Envision Workstation contain everything you need to add fixtures in a laboratory environment. Assemblies include a fine control needle valve service turret with shank, a ¼” reinforced PVC service house, and a male quick connect fitting, which has 316 stainless steel internal parts. Assemblies are designed to only couple with the same key, which is conveniently color coded in orange, yellow, and brown.
Both air and nitrogen quick connects feature a double ended shut off plugs and an internal valve that shuts off when it becomes uncoupled. Vacuum quick connect has a single end shut off plug. The service hoses connects from the overhead service panel through the upper right of the table to the shank of the fixture to provide services. The needle valve features a 135° turret with a polished chrome valve with a brass floating cone and integral brass seat, and can control gases up to 60 psi.
Plumbing assemblies ship in two major components to be installed on site.
Two year warranty
Specifications
  • Material316 stainless steel quick connect, PVC hose, chrome valve with brass
  • ColorOrange
  • Compatible WithEnvision workstations
  • ConnectionsLeft side
  • DescriptionEnvision Complete Plumbing Assemblies, Left Side, Air, Orange
  • Warranty2 Years