ICI Quick Ship Envision Complete Plumbing Assemblies
Plumbing assemblies designed specifically for Envision workstations
  • Conveniently color labeled for easy coupling
  • Control gases up to 60 psi
  • Consist of two major components
  • Available for air, vacuum, and nitrogen services
  • Choose between left and right side assemblies
Plumbing assemblies for the Envision Workstation contain everything you need to add fixtures in a laboratory environment. Assemblies include a fine control needle valve service turret with shank, a ¼” reinforced PVC service house, and a male quick connect fitting, which has 316 stainless steel internal parts. Assemblies are designed to only couple with the same key, which is conveniently color coded in orange, yellow, and brown.
Both air and nitrogen quick connects feature a double ended shut off plugs and an internal valve that shuts off when it becomes uncoupled. Vacuum quick connect has a single end shut off plug. The service hoses connects from the overhead service panel through the upper right of the table to the shank of the fixture to provide services. The needle valve features a 135° turret with a polished chrome valve with a brass floating cone and integral brass seat, and can control gases up to 60 psi.
Plumbing assemblies ship in two major components to be installed on site.
$918.00USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Color Compatible With Description
Availability Pricing
ICI Quick Ship Envision Complete Plumbing Assemblies, Left Side, Air, Orange EW-78984-38 Mfr # CPEISV-A-L
COMPARE
Orange Envision workstations Envision Complete Plumbing Assemblies, Left Side, Air, Orange
$918.00USD / Each
ICI Quick Ship Envision Complete Plumbing Assemblies, Left Side, Vacuum, Yellow EW-78984-39 Mfr # CPEISV-V-L
COMPARE
Yellow Envision workstations Envision Complete Plumbing Assemblies, Left Side, Vacuum, Yellow
$918.00USD / Each
ICI Quick Ship Envision Complete Plumbing Assemblies, Left Side, Nitrogen, Brown EW-78984-40 Mfr # CPEISV-N-L
COMPARE
Brown Envision workstations Envision Complete Plumbing Assemblies, Left Side, Nitrogen, Brown
$918.00USD / Each
ICI Quick Ship Envision Complete Plumbing Assemblies, Right Side, Air, Orange EW-78984-41 Mfr # CPEISV-A-R
COMPARE
Orange Envision workstations Envision Complete Plumbing Assemblies, Right Side, Air, Orange
$918.00USD / Each
ICI Quick Ship Envision Complete Plumbing Assemblies, Right Side, Vacuum, Yellow EW-78984-42 Mfr # CPEISV-V-R
COMPARE
Yellow Envision workstations Envision Complete Plumbing Assemblies, Right Side, Vacuum, Yellow
$918.00USD / Each
ICI Quick Ship Envision Complete Plumbing Assemblies, Right Side, Nitrogen, Brown EW-78984-43 Mfr # CPEISV-N-R
COMPARE
Brown Envision workstations Envision Complete Plumbing Assemblies, Right Side, Nitrogen, Brown
$918.00USD / Each