Imada EX-2 Durometer Hardness Tester for Normal Rubber, Shore A
Representative image only.
Imada EX-2 Durometer Hardness Tester for Normal Rubber, Shore A
Imada | Mfr: E2-A| Item: UX-59792-35
Won’t destroy your sample
$462.80USD / Each
for an additional
$170.25USD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$462.80USD / Each
Buy It Together
Current Item
Imada EX-2 Durometer Hardness Tester for Normal Rubber, Shore A
Imada EX-2 Durometer Hardness Tester for Normal Rubber, Shore A
5979235
$462.80USD / Each
+
InnoCal NIST-Traceable Calibration; Durometer
1708018
$170.25USD / Each
+ + =
Total Price 3068.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Heavy-duty units feature well-made design with high repeatability
  • Measurement scale is 100 points with an accuracy of ±1 point
  • 2 year warranty
More About this Item
Durometers feature portable design, ease of operation, and non-destructive measurement. They are heavy-duty and well-made with high repeatability. The measurement of hardness is an essential factor for determining the characteristics of various rubber, elastomers and plastic products. One of the most important steps in using a hardness tester is to select the optimum type from the various types available based upon different material hardnesses. The measurement scale is 100 points with an accuracy of ± 1 point.
Hardness Tester Selection
  • Model D: Hard rubber and plastics
  • Model A: Normal rubber and softer plastics
  • Asker C: Soft rubber and sponge
Two year warranty
Specifications
  • TypeShore A