LabChem Congo Red Indicator, 0.1% (w/v), pH 3.0-5.0 Blue to Red; 500 mL

Representative image only.

LabChem Congo Red Indicator, 0.1% (w/v), pH 3.0-5.0 Blue to Red; 500 mL

LabChem | Mfr: LC133601| Item: EW-80015-35

Highest quality raw materials!

$59.10USD / Each

$59.10USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity Grade
  • Element/IonpH
  • Volume (mL)500
  • PackagingPoly Bottle
  • Shelf Life (days)730
  • Shelf Life (months)24
  • DescriptionCongo Red Indicator, 0.1% (w/v), pH 3.0-5.0 Blue to Red; 500 mL
Documents & Certificates
Reviews
Q&A