LabChem Congo Red Indicator, 0.1%
Highest quality raw materials!
  • Reagents and solutions to meet your lab and research needs
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$22.60 - $59.10USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Volume (mL) Description
Availability Pricing
LabChem Congo Red Indicator, 0.1% (w/v), pH 3.0-5.0 Blue to Red; 500 mL EW-80015-35 Mfr # LC133601
COMPARE
500 Congo Red Indicator, 0.1% (w/v), pH 3.0-5.0 Blue to Red; 500 mL
$59.10USD / Each
LabChem Congo Red Indicator, 0.1% (w/v), pH 3.0-5.0 Blue to Red; 125 mL EW-80015-37 Mfr # LC133607
COMPARE
125 Congo Red Indicator, 0.1% (w/v), pH 3.0-5.0 Blue to Red; 125 mL
$22.60USD / Each