LabChem Glucose-Glutamic Acid for BOD, Sterilized; 1 L

Representative image only.

LabChem Glucose-Glutamic Acid for BOD, Sterilized; 1 L

LabChem | Mfr: LC148452| Item: EW-80018-09

Highest quality raw materials!

$115.30USD / Each

$115.30USD / Each

Buy It Together

Current Item

LabChem Glucose-Glutamic Acid for BOD, Sterilized; 1 L
LabChem Glucose-Glutamic Acid for BOD, Sterilized; 1 L
8001809

$115.30USD / Each
+ =
Total Price 165.60 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • RatingAPHA 5210 B
  • Purity Grade
  • Element/IonBOD
  • Volume (mL)1000
  • PackagingAmber Glass Bottle
  • Shelf Life (days)730
  • Shelf Life (months)24
  • DescriptionGlucose-Glutamic Acid, for BOD (Sterilized); 1 L
Reviews
Q&A