LabChem Phenolphthalein, 1% In 60% Isopropanol, pH 8.3-10.0 Colorless to Pink; 125 mL

Representative image only.

LabChem Phenolphthalein, 1% In 60% Isopropanol, pH 8.3-10.0 Colorless to Pink; 125 mL

LabChem | Mfr: LC182107| Item: EW-80030-99

Highest quality raw materials!

$22.20USD / Each

$22.20USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity Grade
  • MediumIsopropanol
  • Element/IonpH
  • Volume (mL)125
  • PackagingPoly Bottle
  • DOT InformationClass 3, Packing Group II
  • UN NumberUN1219
  • Shelf Life (days)730
  • Shelf Life (months)24
  • DescriptionPhenolphthalein, 1% In 60% Isopropanol, pH 8.3-10.0 Colorless to Pink; 125 mL
Reviews
Q&A