LabChem Phenolphthalein, 1% In 60% Isopropanol
Highest quality raw materials!
  • Reagents and solutions to meet your lab and research needs
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$22.20 - $69.30USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Volume (mL) Description
Availability Pricing
LabChem Phenolphthalein, 1% In 60% Isopropanol, pH 8.3-10.0 Colorless to Pink; 500 mL EW-80030-72 Mfr # LC182101
COMPARE
500 Phenolphthalein, 1% In 60% Isopropanol, pH 8.3-10.0 Colorless to Pink; 500 mL
$44.60USD / Each
LabChem Phenolphthalein, 1% In 60% Isopropanol, pH 8.3-10.0 Colorless to Pink; 1 L EW-80030-73 Mfr # LC182102
COMPARE
1000 Phenolphthalein, 1% In 60% Isopropanol, pH 8.3-10.0 Colorless to Pink; 1 L
$69.30USD / Each
LabChem Phenolphthalein, 1% In 60% Isopropanol, pH 8.3-10.0 Colorless to Pink; 125 mL EW-80030-99 Mfr # LC182107
COMPARE
125 Phenolphthalein, 1% In 60% Isopropanol, pH 8.3-10.0 Colorless to Pink; 125 mL
$22.20USD / Each