Labconco Protector Premier 4' Fume Hood With Built-in Blower, No Services; 115V/60Hz

Representative image only.

Labconco Protector Premier 4' Fume Hood With Built-in Blower, No Services; 115V/60Hz

Labconco | Mfr: 100400040| Item: EW-09102-63

Excellent chemical resistance

$14,337.00USD / Each
$15,930.00USD / Each

$14,337.00USD / Each
$15,930.00USD / Each

Buy It Together

Current Item

Labconco Protector Premier 4' Fume Hood With Built-in Blower, No Services; 115V/60Hz
Labconco Protector Premier 4' Fume Hood With Built-in Blower, No Services; 115V/60Hz
0910263

$14,337.00USD / Each
$15,930.00USD / Each
+ + +
Labconco 2247400 47" Basic Fume Hood, with no blower, 115 VAC/60 Hz
3350300

$5,809.50USD / Each
$6,455.00USD / Each
+ + =
Total Price 57791.50 ADD 6 ITEMS TO CART

Customers Also Bought

Product Description

More About this Item

Labconco Protector Premier Fiberglass Fume Hoods have a one-piece liner of specially-formulated, fiberglass-reinforced polyester, which offers corrosion and fire resistance and easy clean up. There are no seams where deterioration can occur or where contaminants can collect. These general-purpose hoods are ideal for corrosive fumes but should not be used with perchloric acid, or radioactive or biological hazards.

These by-pass hoods feature the largest unobstructed sightline available—37.5” from work surface to header panel. Other features include Eco-Foil™ air foil that corrects natural turbulence while keeping cords out of the way, dry powder-coated steel exterior, safety glass sash, and molded fiberglass exhaust collar.

Hoods are available with a built-in standard fiberglass blower, an explosion-proof fiberglass blower, or without a blower (remote blower sold separately). Standard hoods include T8 fluorescent lighting. Explosion-proof (EP) models include incandescent lighting (bulb not included). EP blowers are UL-listed for Class I, Group D and Class II, Groups E, F and G. All models are ETL listed. 230V models are CE listed.

Additional service fixtures can be added; contact a Cole-Parmer Application Specialist.

ETL Listed
Specifications
  • Hood Width (in)48
  • Blower TypeStandard
  • Blower (hp)1/3
  • LightsFluorescent
  • Switches2
  • Exterior Width (in)48
  • Exterior Height (in)66
  • Exterior Depth (in)31.6875
  • Power (VAC)115
  • Power (Hz)60
  • Shipping Weight (lbs)365
  • Shipping Weight (kg)196
  • Ductwork Diameter (in)10
  • DescriptionFiberglass 48" Fume Hood, No Services, With Blower, 115 V, 60 Hz
Reviews
Q&A