Mettler Toledo Pure pH Electrode; S7
Representative image only.
Mettler Toledo Pure pH Electrode; S7
Mettler Toledo | Mfr: 30248112| Item: EW-35902-00
Expertly-engineered probes to cover a wide range of applications
$650.00USD / Each
$650.00USD / Each
Buy It Together
Current Item
Customers Also Bought
Product Description
Key Features
  • Cable not included⌡LoT membrane for low temperature and ionic strength samples
More About this Item
These ISM® - Intelligent Sensor Management electrodes models store calibration data and automatically transfer the data to your meter. Choose open junction versions for dirty and complex samples. Cables not included with all electrodes. For additional versatility—select the one that connects to your meter.
Select the appropriate sensor cable for your application and meter—BNC, waterproof BNC, DIN, Lemo OO, and wire-only cables available. ⌡Sleeve junction provides fast results and works best in dirty samples. Special low-temperature glass membrane (LoT) is ideal for working with low temperature and low ionic strength samples.
Specifications
  • ApplicationsUltra-pure water/low ionic strength samples
  • Electrode Body MaterialGlass
  • Reference JunctionImmoveable glass sleeve
  • Reference ElectrolyteFRISCOLYT-B®
  • Sensing Glass TypeLoT - Low temperature and low ionic strength applications
  • Min pH (pH)1
  • Max pH (pH)11
  • Min Temperature (° C)0
  • Max Temperature (° C)80
  • ATC Element
  • ConnectorS7
  • Diameter (mm)12
  • Length (mm)120
  • DescriptionpH Electrode; S7