Microflex FFS700-S Nitrile Gloves, Small; 1000/CS
Representative image only.
Microflex FFS700-S Nitrile Gloves, Small; 1000/CS
Microflex | Mfr: FFS700-S| Item: EW-86445-20
Non-stick properties resist tape and adhesives
$397.70USD / Case of 1000
$397.70USD / Case of 1000
Product Description
Key Features
  • Certified to the Emergency Medical Glove Requirements of NFPA 1999
  • Non-stick properties resist tape and adhesives
  • Powder-free to reduce skin irritation
  • Meets acceptable quality level (AQL) of 1.5
More About this Item
The gloves feature a palm thickness of 3.5 mil and a finger thickness of 5.1 mil, ideal for light to mid-duty work. Lightly textured fingers ensure a secure grip in wet and dry conditions. Interior polymer coating assists in quick and easy donning.
The 100% nitrile gloves offer superior comfort and elasticity in a synthetic glove. Unique formulation conforms to the contours of your hands as you move — minimizing unnecessary hand stress and enabling a full range-of-motion. Ideal for use in the food processing, life sciences and healthcare industries.
Standards and certifications include: ASTM D6319, EN 420:2003 + A1:2009, FDA21 CFR 177-2600-US Food Contact Approved, NFPA 1999:2018, and US QSR/FDA 510(k) Medical Examination Grade. Ambidextrous.
Specifications
  • MaterialNitrile
  • SizeSmall
  • Length (in)9 1/2
  • Length (cm)24
  • ColorBlue
  • SterileNo
  • PowderedNo
  • Palm Thickness (mil)3.5
  • DescriptionNitrile Gloves, small, 1000/cs