Microflex USE-880-S Nitrile Gloves, Extended Cuff, Small; 1000/CS
Representative image only.
Microflex USE-880-S Nitrile Gloves, Extended Cuff, Small; 1000/CS
Microflex | Mfr: USE-880-S| Item: EW-86232-29
Extended cuff provides added protection over wrist and forearm
$315.60USD / Case of 1000
$315.60USD / Case of 1000
Product Description
Key Features
  • Medical examination grade protects hands of healthcare professionals
  • Non-stick surface resists tapes and adhesives
  • Powder-free to reduce skin irritation
  • Meets acceptable quality level (AQL) of 1.5
  • Packed 100 gloves per box - more per box than most extended cuff gloves
More About this Item
These exam gloves feature a palm thickness of 3.9 mil, a finger thickness of 5.1 mil, and an extended cuff for expanded protection over your wrist and forearm. Ultra-soft nitrile formulation also provides protection against hazardous substances while offering a fit and feel not common to nitrile gloves. Textured for a secure grip in wet and dry environments.
The thin nitrile gloves offer tactile sensitivity for light to mid-duty work. Ideal for use in the food processing, life sciences, and healthcare industries.
Standards and certifications include ASTM D6319, EN 420:2003 + A1:2009, FDA21 CFR 177-2600-US Food Contact Approved, and US QSR/FDA 510(k) Medical Examination Grade. Ambidextrous.
Conformite Europeenne
Specifications
  • MaterialNitrile
  • SizeSmall
  • Length (in)11 1/2
  • Length (cm)29
  • ColorBlue
  • GripTextured
  • CuffBeaded
  • SterileNo
  • Palm Thickness (mil)3.9
  • PowderedNo
  • DescriptionNitrile gloves, small, extended cuff, 1000/cs