Microflex UltraSense Nitrile Exam Gloves
Extended cuff provides added protection over wrist and forearm
  • Medical examination grade protects hands of healthcare professionals
  • Non-stick surface resists tapes and adhesives
  • Powder-free to reduce skin irritation
  • Meets acceptable quality level (AQL) of 1.5
  • Packed 100 gloves per box - more per box than most extended cuff gloves
These exam gloves feature a palm thickness of 3.9 mil, a finger thickness of 5.1 mil, and an extended cuff for expanded protection over your wrist and forearm. Ultra-soft nitrile formulation also provides protection against hazardous substances while offering a fit and feel not common to nitrile gloves. Textured for a secure grip in wet and dry environments.
The thin nitrile gloves offer tactile sensitivity for light to mid-duty work. Ideal for use in the food processing, life sciences, and healthcare industries.
Standards and certifications include ASTM D6319, EN 420:2003 + A1:2009, FDA21 CFR 177-2600-US Food Contact Approved, and US QSR/FDA 510(k) Medical Examination Grade. Ambidextrous.
$313.30 - $543.60USD / Case of 1000
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Palm Thickness (mil) Size
Availability Pricing
Microflex USE-880-S Nitrile Gloves, Extended Cuff, Small; 1000/CS EW-86232-29 Mfr # USE-880-S
COMPARE
11 1/2 3.9 Small
$315.60USD / Case of 1000
Microflex USE-880-M Nitrile Gloves, Extended Cuff, Medium; 1000/CS EW-86232-39 Mfr # USE-880-M
COMPARE
11 1/2 3.9 Medium
$543.60USD / Case of 1000
Microflex USE-880-L Nitrile Gloves, Extended Cuff, Large; 1000/CS EW-86232-49 Mfr # USE-880-L
COMPARE
11 1/2 3.9 Large
$314.20USD / Case of 1000
Microflex USE-880-XL Nitrile Gloves, Extended Cuff,  X-Large; 1000/CS EW-86232-59 Mfr # USE-880-XL
COMPARE
11 1/2 3.9 X-Large
$313.30USD / Case of 1000