North Sciences PR Series Lab/Pharmacy Refrigerator, 2°C to 8°C, 395 L, 220 V

Representative image only.

North Sciences PR Series Lab/Pharmacy Refrigerator, 2°C to 8°C, 395 L, 220 V

North Sciences | Mfr: PR-0400-EU| Item: EW-44260-99

Biopharmaceutical refrigerators with precision temperature control and cloud-based monitoring for total sample security

$3,580.00USD / Each

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$3,580.00USD / Each

Customers Also Bought

Product Description

Key Features

  • Features include leveling feet, interior shelves, and rear stoppers Forced-air circulation with special air ducts guarantees inner temperature consistency Unique evaporator design results in better uniformity with no ice build-up

More About this Item

The PR Series high-performance, biopharmaceutical refrigerators offer superior temperature uniformity and outstanding reliability. They are designed to store pharmaceuticals, medicine, vaccines, biological materials, testing reagents, and laboratory materials. The controller allows the inner temperature to be adjusted by increments of 0.1° with multiple sensors to display and accurately control the temperature. A safety door lock prevents unauthorized access. Auto defrost function offers reliable performance in high ambient temperature and high humidity areas.

1 Year Warranty
Specifications
  • Refrigerator Capacity (cu ft)14
  • Refrigerator Capacity (Liters)395
  • Min Temperature - Refrigerator (° F)36
  • Max Temperature - Refrigerator (° F)46.5
  • Min Temperature - Refrigerator (° C)2
  • Max Temperature - Refrigerator (° C)8
  • Temperature Accuracy (° F)32.18
  • Temperature Accuracy (° C)0.1
  • Temperature Control0.1
  • Refrigerator Interior Width (in)20
  • Refrigerator Interior Height (in)61
  • Refrigerator Interior Depth (in)20
  • Refrigerator Interior Width (cm)50
  • Refrigerator Interior Height (cm)155
  • Refrigerator Interior Depth (cm)51
  • Exterior Width (in)24
  • Exterior Height (in)70
  • Exterior Depth (in)26
  • Exterior Width (cm)60
  • Exterior Height (cm)178.5
  • Exterior Depth (cm)65
  • Number of Doors1
  • Number of Shelves6
  • AlarmYes
  • Battery Backup1 Lead-acid (included)
  • Cabinet ConstructionSteel
  • DisplayLCD
  • Power (VAC)220
  • Power (Hz)50
  • Power (amps)1.8
  • Power (watts)175
  • DescriptionLab/Pharmacy Refrigerator, 2°C to 8°C, 395 L, with TraceableLive®, 220 V
Reviews
Q&A