NSI Lab Solutions 775H-10 Enterococcus High Range NCTC 775 Environmental Culture; 10/PK
Representative image only.
NSI Lab Solutions 775H-10 Enterococcus High Range NCTC 775 Environmental Culture; 10/PK
NSI Lab Solutions | Item: UX-05002-42
A single pellet contains a pure culture that will yield a specific certified activity level—activity level is stated on the product lot COA
$175.00USD / Pkg of 10
$175.00USD / Pkg of 10
Product Description
Key Features
  • Hydration fluid dissolves culture pellet in less than a minute
  • No dilution needed—hydrate then directly inoculate media
  • Less than 10% RSD lot precision
  • No more than two passages from primary strain culture
  • ISO 17034 accredited
  • No royalty fee
  • Pack of 10
More About this Item
These microbiological reference materials are designed for use as quantitative control for environmental water analysis. To use, simply transfer the pellet to a sterile room temperature hydration fluid. The culture pellet lyophilized in Microgel-Flash™ matrix dissolves almost instantly with minimum agitation and no incubation. Directly inoculate media without serial dilution for convenience.
Use E. faecalis NCTC 775 culture as a quantitative control for Enterococci and fecal Streptococci analyses. The Certificate of Analysis (CoA) lists value assignments for MPN and plate count methods for total coliform. Store in the freezer.
Download Certificate of Analysis at nsilabsolutions.com/coa-downloads/.
NCTC reference cultures obtained from the National Collection of Type Cultures.
Specifications
  • OrganismE. faecalis
  • Packaging10/PK
  • Product TypeEnvironmental Single Cultures
  • Shelf Life Months (months)18
  • Shipping requirementFrozen ice packs
  • Storage requirementStore between –20 and –10°C
  • StrainNCTC 775, ATCC 19433
  • DescriptionEnterococcus High Range NCTC 775 Environmental Culture; 10/Pk