NSI Lab Solutions Microbiological Environmental Single Cultures
A single pellet contains a pure culture that will yield a specific certified activity level—activity level is stated on the product lot COA
  • Hydration fluid dissolves culture pellet in less than a minute
  • No dilution needed—hydrate then directly inoculate media
  • Less than 10% RSD lot precision
  • No more than two passages from primary strain culture
  • ISO 17034 accredited
  • No royalty fee
These microbiological reference materials are designed for use as quantitative control for environmental water analysis. To use, simply transfer the pellet to a sterile room temperature hydration fluid. The culture pellet lyophilized in Microgel-Flash™ matrix dissolves almost instantly with minimum agitation and no incubation. Directly inoculate media without serial dilution for convenience.
$175.00 - $259.00USD / Pkg of 10
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Organism Packaging Strain
Availability Pricing
COMPARE
E. aerogenes 20/PK NCTC 10006, ATCC 13048
$259.00USD / Pkg of 20
COMPARE
E. coli 10/PK NCTC 9001, ATCC 1175
$175.00USD / Pkg of 10
COMPARE
E. coli 20/PK NCTC 9001, ATCC 1175
$259.00USD / Pkg of 20
COMPARE
E. faecalis 10/PK NCTC 775, ATCC 19433
$175.00USD / Pkg of 10
COMPARE
E. faecalis 20/PK NCTC 775, ATCC 19433
$259.00USD / Pkg of 20
COMPARE
HPC 10/PK NCTC 775
$175.00USD / Pkg of 10
COMPARE
HPC 20/PK NCTC 775
$259.00USD / Pkg of 20
COMPARE
Klebsiella spp 10/PK NCTC 08167
$175.00USD / Pkg of 10
COMPARE
Klebsiella spp 20/PK NCTC 08167
$259.00USD / Pkg of 20
COMPARE
P. aeruginosa 10/PK NCTC 12951
$175.00USD / Pkg of 10
COMPARE
E. aerogenes 10/PK NCTC 10006, ATCC 13048
$175.00USD / Pkg of 10
Showing 1- 11 of 11